missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SYCE1L (aa 47-85) Control Fragment Recombinant Protein

Catalog No. RP107512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107512 Supplier Invitrogen™ Supplier No. RP107512

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66859 (PA5-66859. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in meiosis.

Specifications

Accession Number A8MT33
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 100130958
Name Human SYCE1L (aa 47-85) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias meiosis-related protein; MRP2; SYCE1L; synaptonemal complex central element protein 1 like; synaptonemal complex central element protein 1-like
Common Name SYCE1L
Gene Symbol SYCE1L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QIEDLISRINDLQQAKKKSSEELRETHSLWEALHRELDS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less