missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SV2C (aa 91-139) Control Fragment Recombinant Protein

Catalog No. RP96839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96839 Supplier Invitrogen™ Supplier No. RP96839

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59290 (PA5-59290. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.

Specifications

Accession Number Q496J9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 22987
Name Human SV2C (aa 91-139) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4930527L09Rik; AI841971; KIAA1054; SV2C; Synaptic vesicle glycoprotein 2 C; synaptic vesicle protein 2 C
Common Name SV2C
Gene Symbol SV2C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GEYQGIPSMNQAKDSIVSVGQPKGDEYKDRRELESERRADEEELAQQYE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less