missing translation for 'onlineSavingsMsg'
Learn More

Human STAT5b Alexa Fluor 647-conjugated Antibody, R&D Systems™

Catalog No. IC1584R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1584R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC1584R100 Supplier Bio-Techne Supplier No. IC1584R100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen STAT5b
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 389215
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_036580
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Gene Symbols STAT5B
Host Species Mouse
Immunogen Human STAT5b synthetic peptide MDSQWIPHAQS Accession # NP_036580
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6777
Test Specificity Detects human STAT5b in Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)