missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ST5 (aa 463-562) Control Fragment Recombinant Protein

Catalog No. RP97437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97437 Supplier Invitrogen™ Supplier No. RP97437

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61331. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene was identified by its ability to suppress the tumorigenicity of Hela cells in nude mice. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified.

Specifications

Accession Number P78524
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6764
Name Human ST5 (aa 463-562) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2010004M01Rik; 2610305K15Rik; DENN domain-containing protein 2B; DENN/MADD domain containing 2B; DENND2B; HeLa tumor suppression 1; HTS1; p126; p70; St5; suppression of tumorigenicity 5; suppression of tumorigenicity 5 protein
Common Name ST5
Gene Symbol DENND2B
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSSPTENGTENQPKFGSKSTLEENAYEDIVGDLPKENPYEDVDLKSRRAGRKSQQLSENSLDSLHRMWSPQDRKYNSPPTQLSLKPNSQSLRSGNWSERK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less