missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD31/PECAM (Soluble) ELISA Kit

ELISA

$858.00 - $5,960.00

Specifications

Accession Number P16284
Assay Range 0.47 to 30.0 ng/mL
Assay Sensitivity 0.06 ng/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sPECAM-1
HRP-Conjugate anti-human sPECAM-1 monoclonal antibody
Human sPECAM-1 Standard, 60ng/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5018096
View Documents
Invitrogen™
BMS229
96 Tests N/A
Only null left
 
5018098
View Documents
Invitrogen™
BMS229TEN
10 x 96 Tests N/A
Only null left
 
Description

Description

The Human Soluble CD31/Platelet Endothelial Cell Adhesion Molecule(Hu sCD31/PECAM) ELISA quantitates Hu sCD31/PECAM in human serum, plasma, cell culture supernatants, urine, or other body fluids. The assay will exclusively recognize both natural and recombinant Hu sCD31/PECAM. Principle of the method The Human sCD31/PECAM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
Specifications

Specifications

P16284
0.06 ng/mL
ELISA
Human
Colorimetric Microplate Reader
CD31,CD31/EndoCAM,GPIIA',PECA1,PECAM-1,endoCAM
1.7
HRP
Plasma 10 μL, Serum 10 μL, Supernatant 10 μL, Urine 10 μL
Leukocyte surface protein, CD31, EndoCAM, HEC7
1 hrs 20 mins
0.47 to 30.0 ng/mL
HRP
Plasma, Serum, Supernatant, Urine
ELISA
5175
7.4%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
RUO
2°C to 8°C
CD31 (PECAM-1)
3 hrs 10 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'