missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPATS2L (aa 139-217) Control Fragment Recombinant Protein

Catalog No. RP101589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101589 Supplier Invitrogen™ Supplier No. RP101589

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63127. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SPATS2L is a protein coding gene.

Specifications

Accession Number Q9NUQ6
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 26010
Name Human SPATS2L (aa 139-217) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2810022L02Rik; A230104H11Rik; AA987173; AI480531; AI842453; AW413586; DNA polymerase transactivated protein 6; DNA polymerase-transactivated protein 6; DNAPTP6; RGD1309930; SGNP; SP1224; Spats2l; SPATS2-like protein; spermatogenesis associated serine rich 2 like; spermatogenesis associated, serine rich 2 like; spermatogenesis associated, serine-rich 2-like; stress granule and nucleolar protein
Common Name SPATS2L
Gene Symbol SPATS2L
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EPSKALRGVTEGNRLLQQKLSLDGNPKPIHGTTERSDGLQWSAEQPCNPSKPKAKTSPVKSNTPAAHLEIKPDELAKKR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less