missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPACA9 (aa 152-222) Control Fragment Recombinant Protein

Catalog No. RP104927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104927 Supplier Invitrogen™ Supplier No. RP104927
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (32%), Rat (32%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65674 (PA5-65674. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SPACA9 (Sperm Acrosome Associated 9) is a Protein Coding gene.

Specifications

Accession Number Q96E40
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11092
Name Human SPACA9 (aa 152-222) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700026L06Rik; 80-kDa SOGA fragment; Acrosome and sperm tail protein; C11H9orf9; C20orf117; C9H9orf9; C9orf9; ci009; C-terminal 80 kDa form; KIAA0889; MAST; N-terminal form; Protein SOGA1; RIKEN cDNA 1700026L06 gene; Rsb66; Rsb66 homolog; Rsb-66 protein; SOGA; SOGA family member 1; SOGA1; SPACA9; sperm acrosome associated 9; sperm acrosome-associated protein 9; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1; uncharacterized protein C9orf9 homolog
Common Name SPACA9
Gene Symbol SPACA9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IAHSEKLPRKVLQHVSEPQAHQESTRGAARPAQAIGTQPRATKHKCRQLTKASLKPRGCSKPPWRPPGGKL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less