missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SOX5 (aa 39-130) Control Fragment Recombinant Protein

Catalog No. RP105155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105155 Supplier Invitrogen™ Supplier No. RP105155
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66331 (PA5-66331. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Accession Number P35711
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6660
Name Human SOX5 (aa 39-130) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A730017D01Rik; AI528773; LAMSHF; L-SO x 5; L-SO x 5 B; L-SO x 5 F; MGC35153; So x 5; Sox-5; So x 5-BLM isoform; So x 5l1; SRY (sex determining region Y)-box 5; SRY box 5; SRY-box 5; SRY-box containing gene 5; SRY-box containing gene 5-like 1; SRY-box transcription factor 5; SRY-related HMG-box 5 protein; transcription factor SOX-5
Common Name SOX5
Gene Symbol SOX5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VEEEESDGLPAFHLPLHVSFPNKPHSEEFQPVSLLTQETCGHRTPTSQHNTMEVDGNKVMSSFAPHNSSTSPQKAEEGGRQSGESLSSTALG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less