missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SNX2 (aa 77-149) Control Fragment Recombinant Protein

Catalog No. RP96984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96984 Supplier Invitrogen™ Supplier No. RP96984
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83367 (PA5-83367. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in several stages of intracellular trafficking.

Specifications

Accession Number O60749
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6643
Name Human SNX2 (aa 77-149) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610030A03Rik; CTB-36H16.2; hypothetical protein LOC509769; MGC5204; SN x 2; sorting nexin 2; sorting nexin-2; Transformation-related gene 9 protein; TRG9; TRG-9
Common Name SNX2
Gene Symbol SNX2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ATEEVSLDSPEREPILSSEPSPAVTPVTPTTLIAPRIESKSMSAPVIFDRSREEIEEEANGDIFDIEIGVSDP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less