missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SNX2 (aa 38-73) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP109875
Description
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145171 (PA5-145171. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in several stages of intracellular trafficking.Specifications
| O60749 | |
| Blocking Assay, Control | |
| 6643 | |
| 100 μL | |
| 0610030A03Rik; CTB-36H16.2; hypothetical protein LOC509769; MGC5204; SN x 2; sorting nexin 2; sorting nexin-2; Transformation-related gene 9 protein; TRG9; TRG-9 | |
| SNX2 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human SNX2 (aa 38-73) Control Fragment | |
| RUO | |
| SNX2 | |
| Unconjugated | |
| Recombinant | |
| SPSSPEPASLPAEDISANSNGPKPTEVVLDDDREDL | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |