missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLURP1 (aa 29-100) Control Fragment Recombinant Protein

Catalog No. RP98422
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98422 Supplier Invitrogen™ Supplier No. RP98422
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62280 (PA5-62280. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number P55000
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57152
Name Human SLURP1 (aa 29-100) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias anti-neoplastic urinary protein; ANUP; ARS; ARS component B; ARS(component B)-81/S; ArsB; LY6LS; lymphocyte antigen 6-like secreted; MDM; secreted LY6/PLAUR domain containing 1; secreted Ly6/uPAR related protein 1; secreted Ly-6/uPAR-related protein 1; SLURP1; SLURP-1
Common Name SLURP1
Gene Symbol SLURP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KEPMTSASCRTITRCKPEDTACMTTLVTVEAEYPFNQSPVVTRSCSSSCVATDPDSIGAAHLIFCCFRDLCN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less