missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC52A3 (aa 243-292) Control Fragment Recombinant Protein

Catalog No. RP99331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99331 Supplier Invitrogen™ Supplier No. RP99331
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61954 (PA5-61954. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a riboflavin transporter protein that is strongly expressed in the intestine and likely plays a role in intestinal absorption of riboflavin. The protein is predicted to have eleven transmembrane domains and a cell surface localization signal in the C-terminus. Mutations at this locus have been associated with Brown-Vialetto-Van Laere syndrome and Fazio-Londe disease. [provided by RefSeq, Mar 2012].

Specifications

Accession Number Q9NQ40
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 113278
Name Human SLC52A3 (aa 243-292) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310046K01Rik; bA371L19.1; BVVLS; BVVLS1; C20orf54; ct054; hRFT2; Rft2; RFVT3; RGD1304644; Riboflavin transporter 2; rRFT2; Slc52a3; solute carrier family 52 (riboflavin transporter), member 3; solute carrier family 52 member 3; solute carrier family 52, riboflavin transporter, member 3; solute carrier protein family 52, member 3
Common Name SLC52A3
Gene Symbol SLC52A3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RQPRCWEASVEDLLNDQVTLHSIRPREENDLGPAGTVDSSQGQGYLEEKA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less