missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC36A2 (aa 11-57) Control Fragment Recombinant Protein

Catalog No. RP96963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96963 Supplier Invitrogen™ Supplier No. RP96963
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (53%), Rat (53%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60580 (PA5-60580. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

regulates pH-dependent, Na(+)-independent uptake of glycine, proline, and alpha(methyl)aminoisobutyric acid.

Specifications

Accession Number Q495M3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 153201
Name Human SLC36A2 (aa 11-57) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias FLJ16051; MGC119658; MGC119660; PAT2; Proton/amino acid transporter 2; Proton-coupled amino acid transporter 2; SLC36A2; solute carrier family 36 (proton/amino acid symporter), member 2; solute carrier family 36 member 2; TRAMD1; tramdorin-1
Common Name SLC36A2
Gene Symbol SLC36A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QGAVAIKLDLMSPPESAKKLENKDSTFLDESPSESAGLKKTKGITVF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less