missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC22A25 (aa 516-547) Control Fragment Recombinant Protein

Catalog No. RP97548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97548 Supplier Invitrogen™ Supplier No. RP97548
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58770 (PA5-58770. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLC22A25 gene ontology annotations related to this gene include transmembrane transport.

Specifications

Accession Number Q6T423
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 387601
Name Human SLC22A25 (aa 516-547) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HIMTP; integral membrane transport UST1r; MGI:2442751, MGI:2385316, MGI:3042283, MGI:3645714, MGI:3605624, MGI:2442750; organic anion transporter UST6; putative integral membrane transport UST1r; putative UST1-like organic anion transporter; SLC22A25; Slc22a9; solute carrier family 22 (organic anion/cation transporter), member 9; solute carrier family 22 member 24; solute carrier family 22 member 25; solute carrier family 22, member 25; Ust1; Ust1r; UST6
Common Name SLC22A25
Gene Symbol SLC22A25
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PETRNQPLLDSIQDVENEGVNSLAAPQRSSVL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less