missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC19A2 (aa 27-92) Control Fragment Recombinant Protein

Catalog No. RP89150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89150 Supplier Invitrogen™ Supplier No. RP89150

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53456 (PA5-53456. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLC19A2 encodes a thiamin transporter protein. Mutations in this gene cause thiamin-responsive megaloblastic anemia syndrome (TRMA), which is an autosomal recessive disorder characterized by diabetes mellitus, megaloblastic anemia and sensorineural deafness.

Specifications

Accession Number O60779
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10560
Name Human SLC19A2 (aa 27-92) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AV276020; AW322295; DDA1; high affinity thiamine transporter; reduced folate carrier protein (RFC) like; Slc19a2; solute carrier family 19 (thiamine transporter), member 2; solute carrier family 19 member 2; TC1; Thiamine carrier 1; Thiamine transporter 1; THMD1; THT1; THTR1; ThTr-1; TRMA
Common Name SLC19A2
Gene Symbol SLC19A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RECWFLPTALLCAYGFFASLRPSEPFLTPYLLGPDKNLTEREEPKPDRLLVLKVLWNDFLMCYSSR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less