missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC18B1 (aa 1-31) Control Fragment Recombinant Protein

Catalog No. RP104880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104880 Supplier Invitrogen™ Supplier No. RP104880

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65574 (PA5-65574. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein, which has high sequence similarity to rat, xenopus and zebrafish proteins. The protein function is unknown.

Specifications

Accession Number Q6NT16
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 116843
Name Human SLC18B1 (aa 1-31) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110021L09Rik; C6orf192; dJ55C23.6; MFS-type transporter C6orf192; MFS-type transporter C6orf192 homolog; MFS-type transporter SLC18B1; RGD1306962; SLC18B1; Solute carrier family 18 member B1; solute carrier family 18, subfamily B, member 1
Common Name SLC18B1
Gene Symbol SLC18B1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MEALGDLEGPRAPGGDDPAGSAGETPGWLSR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less