missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SKIV2L (aa 493-571) Control Fragment Recombinant Protein

Catalog No. RP99551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99551 Supplier Invitrogen™ Supplier No. RP99551

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-140068 (PA5-140068, PA5-62966 (PA5-62966. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.

Specifications

Accession Number Q15477
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6499
Name Human SKIV2L (aa 493-571) Control Fragment
Quantity 100 μL
Gene Alias 170 A; 4930534J06Rik; AW214248; Dd x 13; Helicase SKI2W; helicase-like protein; HLP; SKI; SKI2; SKI2 homolog, superkiller viralicidic activity 2-like; Ski2 like RNA helicase; Ski2w; SKIV2; SKIV2L; SKIV2L1; superkiller viralicidic activity 2-like; superkiller viralicidic activity 2-like (S. cerevisiae); THES2; W
Common Name SKIV2L
Gene Symbol SKIV2L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TGNSSKTQGELFLLLDSRGAFHTKGYYAAVEAKKERMSKHAQTFGAKQPTHQGGPAQDRGVYLSLLASLRTRAQLPVVV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less