missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-3/CD33 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | Siglec-3/CD33 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 996816 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P20138 |
| Gene Alias | CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 |
| Host Species | Mouse |
| Show More |