missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SHQ1 (aa 479-577) Control Fragment Recombinant Protein

Catalog No. RP100146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100146 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' RP100146
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60124 (PA5-60124. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SHQ1 is the human homolog of yeast Shq1p, a protein that acts as an accessory factor in the assembly of H/ACA ribonucleoproteins. SHQ1 has been shown to bind the major H/ACA core protein and the pseudouridine synthase NAP57, but precedes the assembly role of NAF1 and of the other H/ACA core proteins. SHQ1 assists in the assembly of H/ACA-box ribonucleoproteins that function in the processing of ribosomal RNAs, modification of spliceosomal small nuclear RNAs, and stabilization of telomerase.

Specifications

Accession Number Q6PI26
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55164
Name Human SHQ1 (aa 479-577) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2810403P18Rik; FLJ10539; gene associated with retinoid and interferon-induced mortality 1; GRIM-1; hypothetical protein LOC507486; protein SHQ1 homolog; RGD1310610; Shq1; SHQ1 homolog; SHQ1 homolog (S. cerevisiae); SHQ1, H/ACA ribonucleoprotein assembly factor; Shq1p; zgc:158255
Common Name SHQ1
Gene Symbol SHQ1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ELKDSPSETVSSLQGPFLEESSAFLIVDGGVRRNTAIQESDASQGKPLASSWPLGVSGPLIEELGEQLKTTVQVSEPKGTTAVNRSNIQERDGCQTPNN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less