missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SHB (aa 340-400) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP106373

Catalog No. RP106373

Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65701 (PA5-65701. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The SH2 (Src Homology 2) domain is a structurally conserved motif that contains two alpha helices and seven beta strands and is found in a variety of proteins that are involved in signal transduction throughout the cell. Specifically, the SH2 domain targets SH2 domain-containing proteins to tyrosinephosphorylated sites, an event that can trigger a protein-protein interaction cascade which may ultimately effect gene expression and cellular function. Shb (SH2 domain-containing adapter protein b), Shd (SH2 domain-containing adapter protein d), She (SH2 domain-containing adapter protein e) and Shf(SH2 domain-containing adapter protein f) are SH2 domain-containing proteins that play various roles throughout the cell.
Specifications

Specifications

Q15464
Blocking Assay, Control
6461
100 μL
bA3J10.2; BC028832; SH2 domain containing adaptor protein B; SH2 domain-containing adapter protein B; Shb; SHB (Src homology 2 domain containing) adaptor protein B; SHB adaptor protein (a Src homology 2 protein); Src homology 2 domain containing adaptor protein B; src homology 2 domain-containing transforming protein B
SHB
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human SHB (aa 340-400) Control Fragment
RUO
SHB
Unconjugated
Recombinant
WEWNRVTIPALAAQFNGNEKRQSSPSPSRDRRRQLRAPGGGFKPIKHGSPEFCGILGERVD
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions