missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SGTB (aa 285-304) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP103519
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64128 (PA5-64128. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SGTB is co-chaperone that binds directly to HSC70 and HSP70 and regulates their ATPase activity.Specifications
| Q96EQ0 | |
| Blocking Assay, Control | |
| 54557 | |
| 100 μL | |
| Beta-SGT; C630001O05Rik; SGT2; Sgtb; small glutamine rich protein with tetratricopeptide repeats 2; small glutamine rich tetratricopeptide repeat containing beta; small glutamine-rich protein with tetratricopeptide repeats 2; small glutamine-rich tetratricopeptide repeat (TPR)-containing, beta; small glutamine-rich tetratricopeptide repeat-containing protein beta | |
| SGTB | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human SGTB (aa 285-304) Control Fragment | |
| RUO | |
| SGTB | |
| Unconjugated | |
| Recombinant | |
| EQLRNHIRSRSFSSSAEEHS | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |