missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SF3A2 (aa 18-105) Control Fragment Recombinant Protein

Catalog No. RP98921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98921 Supplier Invitrogen™ Supplier No. RP98921

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61969 (PA5-61969. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes subunit 2 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 2 interacts with subunit 1 through its amino-terminus while the single zinc finger domain of subunit 2 plays a role in its binding to the 15S U2 snRNP. Subunit 2 may also function independently of its RNA splicing function as a microtubule-binding protein.

Specifications

Accession Number Q15428
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8175
Name Human SF3A2 (aa 18-105) Control Fragment
Quantity 100 μL
Gene Alias 66 kDa; AW047242; pre-mRNA splicing factor SF3A, subunit 2; PRP11; PRPF11; SAP 62; Sap62; Sf3a2; SF3a66; SFA66; spliceosomal protein gene 62; spliceosome associated protein 62; spliceosome-associated protein 62; Splicing factor 3 A subunit 2; splicing factor 3 A, subunit 2; splicing factor 3 A, subunit 2, 66 kD; splicing factor 3 A, subunit 2, 66 kDa
Common Name SF3A2
Gene Symbol SF3A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SSSESNRDRRERLRQLALETIDINKDPYFMKNHLGSYECKLCLTLHNNEGSYLAHTQGKKHQTNLARRAAKEAKEAPAQPAPEKVKVE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less