missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serum Albumin Monoclonal Antibody (AL-01), Biotin
GREENER_CHOICE

Catalog No. PIMA110051
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110051 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110051 Supplier Invitrogen™ Supplier No. MA110051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

This antibody reacts only with human serum albumin, a 65-67 kDa monomeric protein in human blood plasma; it is produced in liver. No cross-reactivity was observed with other serum proteins.

Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough 'produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.

Specifications

Antigen Human Serum Albumin
Applications ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone AL-01
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 15mM sodium azide; pH 7.4
Gene ALB
Gene Accession No. P02768
Gene Alias Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341
Gene Symbols ALB
Host Species Mouse
Immunogen Fraction of proteins containing albumin after ammonnium sulphate precipitation and DEAE-chromatography of human serum.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less