missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GeminiBio™ Human Serum AB (Off-the-Clot)
Description
- Human Serum AB, Off-the-Clot is designed for cell therapy programs that prioritize biological fidelity, defined sourcing, and regulatory readiness.
- Produced through natural blood coagulation, this serum format preserves native composition while avoiding the use of anticoagulants or preservatives.
- Extensively tested and sourced from a controlled donor pool, supporting clinical and commercial applications where consistency, traceability, and supply continuity matter.
- Naturally coagulated (off-the-clot) serum format with no added anticoagulants or preservatives
- Collected from healthy male donors of the AB serotype
- Extensively tested to support clinical and commercial workflows
- Defined donor pool (>50 donors) balancing traceability with supply continuity
- Clear processing methodology supporting raw material control and consistency expectations
Suited for:
- Clinical and commercial programs seeking a minimally processed Human Serum AB format
- Applications prioritizing native serum characteristics
- Teams requiring established testing, documentation, and long-term supply planning
Specifications
Specifications
| Form | Frozen |
| Format | Bottle |
| Product Type | Human Serum |
| Quantity | 100 mL |
| Sterility | Sterile |
| Sterilization Method | Gamma-irradiated |
| Mycoplasma Detected | No |
| Gamma-irradiated | Yes |