missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Serpin B3/SCCA1 ELISA Kit
ELISA
Supplier: Invitrogen™ EH413RB
Description
Human Serpin B3/SCCA1 quantitates human Serpin B3/SCCA1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin B3/SCCA1.
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt]Chemical Identifiers
| HsT1196; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin B4; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
| 0.15 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| SERPINB3 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| SERPINB3 | |
| 4 hrs 45 mins |
| 0.156 to 10 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 6317 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| HsT1196; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin B4; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
| 0.15 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| SERPINB3 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.156 to 10 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 6317 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| SERPINB3 | |
| 4 hrs 45 mins |