missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Septin 1 (aa 284-336) Control Fragment Recombinant Protein

Catalog No. RP98781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98781 Supplier Invitrogen™ Supplier No. RP98781

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83564 (PA5-83564. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintenance of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.

Specifications

Accession Number Q8WYJ6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1731
Name Human Septin 1 (aa 284-336) Control Fragment
Quantity 100 μL
Gene Alias 1-Sep; DIFF6; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); differentiation protein 6; LARP; MGC20394; Peanut-like protein 3; PNUTL3; Protein Diff6; SEP1; Sept1; septin 1; Septin1; septin-1; Serologically defined breast cancer antigen NY-BR-24
Common Name Septin 1
Gene Symbol SEPTIN1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EVTHDLLYEGYRARCLQSLARPGARDRASRSKLSRQSATEIPLPMLPLADTEK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less