missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Semaphorin 6A ELISA Kit

Catalog No. EH407RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH407RBX5 5 x 96 Tests
EH407RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH407RBX5 Supplier Invitrogen™ Supplier No. EH407RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

ELISA

Human Semaphorin 6A quantitates human Semaphorin 6A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin 6A.

SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.

Specifications

Accession Number Q9H2E6
Assay Range 0.4-100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA;
Gene ID (Entrez) 57556
Gene Symbol SEMA6A
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target SEMA6A
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less