missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 3E Alexa Fluor 405-conjugated Antibody, R&D Systems™
Description
Semaphorin 3E Monoclonal specifically detects Semaphorin 3E in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | Semaphorin 3E |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 400513 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 405 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | O15041 |
| Gene Alias | (semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E |
| Show More |
For Research Use Only (RUO)