missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SCHIP1 Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP108559
Description
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84827 (PA5-84827. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This locus represents naturally occurring read-through transcription from the neighboring IQ motif containing J (IQCJ) and schwannomin interacting protein 1 (SCHIP1) genes. Alternative splicing results in multiple transcript variants that are composed of in-frame exons from each individual gene. The resulting fusion products are thought to be components of the multimolecular complexes of axon initial segments and nodes of Ranvier, and they may play a role in calcium-mediated responses.Specifications
| P0DPB3 | |
| Blocking Assay, Control | |
| 29970 | |
| 100 μL | |
| hypothetical protein LOC550605; LOC478678; MGC128591 protein; neurofibromatosis 2 interacting protein; Nf2ip; Schip1; SCHIP-1; schwannomin interacting protein 1; schwannomin-interacting protein 1; zgc:110585 | |
| Schip1 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human SCHIP1 Control Fragment | |
| RUO | |
| SCHIP1 | |
| Unconjugated | |
| Recombinant | |
| MNLDSDGMDDIISQESSLDMEGNYKKAQKNERES | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |