missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human SCGF-alpha Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $8480.00
Specifications
| Accession Number | Q9Y240 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 6320 |
| Molecular Weight (g/mol) | 33.9 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10022A100UG
|
Gibco™
10022A100UG |
100 μg |
Each for $1,614.00
|
|
|||||
10022A10UG
|
Gibco™
LS10022A10UG |
10 μg |
Each for $318.00
|
|
|||||
10022A1MG
|
Gibco™
LS10022A1MG |
1 mg |
Each for $8,480.00
|
|
|||||
10022A250UG
|
Gibco™
10022A250UG |
250 μg |
Each for $2,825.00
|
|
|||||
10022A2UG
|
Gibco™
LS10022A2UG |
2 μg |
Each for $132.00
|
|
|||||
10022A500UG
|
Gibco™
10022A500UG |
500 μg |
Each for $4,835.00
|
|
|||||
Description
Recombinant Human SCGF-alpha is a 33.9 kDa polypeptide containing 305 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.Specifications
| Q9Y240 | |
| protein with no preservative | |
| 33.9 kDa | |
| E. coli | |
| <1 EU/ μg | |
| CLEC11A | |
| Determined by its inhibitory effect on the proliferation of TF-1 cells previously grown in media containing GM-CSF. The ED50 for this effect was observed (at ≤ 2.5 ng/ml) | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 6320 | |
| Human SCGF-alpha | |
| RUO | |
| AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF | |
| CLEC11A | |
| Unconjugated | |
| GHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ AAAQARCTAR GGSLAQPADR QQMEALTRYL RAALAPYNWP VWLGVHDRRA EGLYLFENGQ RVSFFAWHRS PRPELGAQPS ASPHPLSPDQ PNGGTLENCV AQASDDGSWW DHDCQRRLYY VCEFPF | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |