missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SC5D (aa 57-79) Control Fragment Recombinant Protein

Catalog No. RP105499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105499 Supplier Invitrogen™ Supplier No. RP105499

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67092 (PA5-67092. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Catalyzes a dehydrogenation to introduce C5-6 double bond into lathosterol.

Specifications

Accession Number O75845
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6309
Name Human SC5D (aa 57-79) Control Fragment
Quantity 100 μL
Gene Alias 3 beta-hydroxysteroid-delta5-desaturase; C-5 sterol desaturase; Delta(7)-sterol 5-desaturase; delta(7)-sterol C5(6)-desaturase; ERG3; fungal ERG3, delta-5-desaturase-like; lathosterol 5-desaturase; lathosterol dehydrogenase; lathosterol oxidase; S5DES; SC5D; SC5DL; Sterol-C5-desaturase; sterol-C5-desaturase (ERG3 delta-5-desaturase homolog, S. cerevisiae)-like
Common Name SC5D
Gene Symbol SC5D
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VFDHALMKHPQFLKNQVRREIKF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less