missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SARS-CoV-2 Spike (Trimer) IgG ELISA Kit
ELISA
Supplier: Invitrogen™ BMS2325TEN
Description
It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.Specifications
| 234 to 15,000 Units/mL | |
| HRP | |
| Plasma, Serum | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 30 mins |
| <39 Units/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| <10% | |
| Pre-coated 96 well plate, High Control (standard), Calibrator (medium control), Low Control, IgG - HRP conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers | |
| 10 x 96 Tests | |
| Plasma 5 μL, Serum 5 μL | |
| SARS-CoV-2 Spike (trimer) Protein | |
| 1 hrs 15 mins |