missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SAB (aa 317-403) Control Fragment Recombinant Protein

Catalog No. RP96330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96330 Supplier Invitrogen™ Supplier No. RP96330

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57662 (PA5-57662. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Inhibits the auto- and transphosphorylation activity of BTK. Plays a negative regulatory role in BTK-related cytoplasmic signaling in B-cells. May be involved in BCR-induced apoptotic cell death.

Specifications

Accession Number O60239
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9467
Name Human SAB (aa 317-403) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI606498; Sab; SH3 binding protein; SH3 domain binding protein 5; SH3 domain-binding protein 5; SH3 domain-binding protein that preferentially associates with BTK; SH3BP5; SH3BP-5; SH3-domain binding protein 5 (BTK-associated); Vascular endothelial cell-specific protein 18; Vesp18
Common Name SAB
Gene Symbol SH3BP5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ETQSVSSFSSGPTSPSEMPDQFPAVVRPGSLDLPSPVSLSEFGMMFPVLGPRSECSGASSPECEVERGDRAEGAENKTSDKANNNRG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less