missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100P |
|---|---|
| Assay Range | 14 to 3,500 pg/mL |
| Assay Sensitivity | 14 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH401RBX5
|
Invitrogen™
EH401RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH401RB
|
Invitrogen™
EH401RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human S100 P quantitates human S100 P in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human S100 P.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 4p16. This protein, in addition to binding Ca2+, also binds Zn2+ and Mg2+. This protein may play a role in the etiology of prostate cancer.Specifications
| MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100P | |
| 14 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| S100P | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| S100P | |
| 4 hrs 45 mins |
| 14 to 3,500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 6286 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |