missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human S100 A2 ELISA Kit

Catalog No. EH398RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH398RBX5 5 x 96 Tests
EH398RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH398RBX5 Supplier Invitrogen™ Supplier No. EH398RBX5

ELISA

Human S100 A2 quantitates human S100 A2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human S100 A2.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may have a tumor suppressor function. Chromosomal rearrangements and altered expression of this gene have been implicated in breast cancer.

Specifications

Accession Number CAN19; EG628324; predicted gene, EG628324; Protein S100-A2; Protein S-100 L; S100 calcium binding protein A2; S100 calcium-binding protein A2; S100A2; S100L
Assay Range 0.05 to 10 ng/mL
Assay Sensitivity 0.05 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6273
Gene Symbol S100A2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target S100A2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less