missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RTKN2 (aa 279-365) Control Fragment Recombinant Protein

Catalog No. RP97196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97196 Supplier Invitrogen™ Supplier No. RP97196

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58392 (PA5-58392. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RTKN2 may play an important role in lymphopoiesis.

Specifications

Accession Number Q8IZC4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 219790
Name Human RTKN2 (aa 279-365) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias B130039D23Rik; bA531F24.1; Mbf; membrane bound factor; PH domain-containing family K member 1; pleckstrin homology domain containing, family K member 1; pleckstrin homology domain-containing family K member 1; Plekhk1; rhotekin 2; rhotekin-2; Rtkn2
Common Name RTKN2
Gene Symbol RTKN2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AQPACMAEDAFAGFLNQQQMVEGLISWRRLYCVLRGGKLYCFYSPEEIEAKVEPALVVPINKETRIRAMDKDAKKRIHNFSVINPVP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less