missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ROBO2 (aa 934-1070) Control Fragment Recombinant Protein

Catalog No. RP90546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP90546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP90546 Supplier Invitrogen™ Supplier No. RP90546

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82481. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.

Specifications

Accession Number Q9HCK4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6092
Name Human ROBO2 (aa 934-1070) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549
Common Name ROBO2
Gene Symbol ROBO2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PVNNSNSGPNEIGNFGRGDVLPPVPGQGDKTATMLSDGAIYSSIDFTTKTSYNSSSQITQATPYATTQILHSNSIHELAVDLPDPQWKSSIQQKTDLMGFGYSLPDQNKGNNGGKGGKKKKNKNSSKPQKNNGSTWA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less