missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RILPL1 (aa 82-169) Control Fragment Recombinant Protein

Catalog No. RP97878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97878 Supplier Invitrogen™ Supplier No. RP97878

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59512. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plays a role in the regulation of cell shape and polarity. Plays a role in cellular protein transport, including protein transport away from primary cilia. Neuroprotective protein, which acts by sequestring GAPDH in the cytosol and prevent the apoptotic function of GAPDH in the nucleus. Competes with SIAH1 for binding GAPDH. Does not regulate lysosomal morphology and distribution (PubMed:14668488). Binds to RAB10 following LRRK2-mediated RAB10 phosphorylation which leads to inhibition of ciliogenesis (PubMed:30398148). [UniProt].

Specifications

Accession Number Q5EBL4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 353116
Name Human RILPL1 (aa 82-169) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2900002H16Rik; 6330559I19Rik; GAPDH competitor of SIAH protein enhances life; GAPDH's competitor of SIAH1 protein enhances life; GOSPEL; MNCb-2440; Rab interacting lysosomal protein like 1; Rab interacting lysosomal protein-like 1; rab-interacting lysosomal-like protein 1; RGD1307973; Rilpl1; RILP-like protein 1; RLP1
Common Name RILPL1
Gene Symbol RILPL1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LELDRLRLERMDRIEKERKHQKELELVEDVWRGEAQDLLSQIAQLQEENKQLMTNLSHKDVNFSEEEFQKHEGMSERERQVMKKLKEV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less