missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RGS12 (aa 339-432) Control Fragment Recombinant Protein

Catalog No. RP106559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106559 Supplier Invitrogen™ Supplier No. RP106559

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65886 (PA5-65886. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the 'regulator of G protein signaling' (RGS) gene family. The encoded protein may function as a guanosine triphosphatase (GTPase)-activating protein as well as a transcriptional repressor. This protein may play a role in tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their biological nature has not been determined.

Specifications

Accession Number O14924
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6002
Name Human RGS12 (aa 339-432) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1200016K18Rik; 4632412M04Rik; AI481290; DKFZp761K1617; DKFZp761K1817; E130309H11; regulator of G-protein signaling 12; regulator of G-protein signalling 12; Rgs12; RP11-529E10.1
Common Name RGS12
Gene Symbol RGS12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VFMVDPDLFNHKIHQGIARRFGFECTADPDTNGCLEFPASSLPVLQFISVLYRDMGELIEGMRARAFLDGDADAHQNNSTSSNSDSGIGNFHQE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less