Learn More
Invitrogen™ Human RANKL (Soluble) ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier: Invitrogen™ 900K142K
Description
This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found.Specifications
| O14788 | |
| Lot-specific | |
| ELISA | |
| 8600 | |
| Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
| 10 x 96 Tests | |
| Refer to product documentation for component specific storage temperature. | |
| 1 hrs 45 mins |
| Lot-specific | |
| HRP | |
| Colorimetric Microplate Reader | |
| TNFSF11 | |
| HRP | |
| RUO | |
| RANKL (RANK Ligand) | |
| 24 hrs 30 mins |