missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PVRIG (aa 214-314) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100006
Description
Highest antigen sequence indentity to the following orthologs: Mouse (30%), Rat (30%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61535 (PA5-61535. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Cell surface receptor for NECTIN2. May act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals. Following interaction with NECTIN2, inhibits T-cell proliferation. Competes with CD226 for NECTIN2-binding.Specifications
| Q6DKI7 | |
| Blocking Assay, Control | |
| 79037 | |
| 100 μL | |
| C7orf15; CD112 receptor; CD112R; poliovirus receptor related immunoglobulin domain containing; poliovirus receptor-related immunoglobulin domain-containing protein; PVRIG; Transmembrane protein PVRIG | |
| PVRIG | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human PVRIG (aa 214-314) Control Fragment | |
| RUO | |
| PVRIG | |
| Unconjugated | |
| Recombinant | |
| PRARAWAPSQASQAALHVPYATINTSCRPATLDTAHPHGGPSWWASLPTHAAHRPQGPAAWASTPIPARGSFVSVENGLYAQAGERPPHTGPGLTLFPDPR | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |