missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PUSL1 (aa 232-303) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP105972

Catalog No. RP105972

Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83482 (PA5-83482. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PUSL1 is a protein coding gene. Gene ontology (GO) annotation include intracellular membrane-bounded organelle.
Specifications

Specifications

Q8N0Z8
Blocking Assay, Control
126789
100 μL
pseudouridylate synthase-like 1; PUSL1; tRNA pseudouridine synthase-like 1; tRNA pseudouridylate synthase-like 1; tRNA-uridine isomerase-like 1
PUSL1
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human PUSL1 (aa 232-303) Control Fragment
RUO
PUSL1
Unconjugated
Recombinant
RQVRRMTAVLVAVGLGALAPAQVKTILESQDPLGKHQTRVAPAHGLFLKSVLYGNLGAASCTLQGPQFGSHG
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions