missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PTPRCAP (aa 112-169) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100004
Description
Highest antigen sequence indentity to the following orthologs: Mouse (33%), Rat (33%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60487 (PA5-60487. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MSK-1 is a mitogen and stress activated protein kinase-1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK-1 can be activated by ERK1/2 and SAPK2 /p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG-14 in response to mitogen and stress. Similar to RSK, MSK-1 contains two kinase domains (N-term and a C-term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK-1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C-terminal kinase domain of MSK-1 which is essential for the catalytic activity of the N-terminal kinase domain.Specifications
| Q14761 | |
| Blocking Assay, Control | |
| 5790 | |
| 100 μL | |
| AI605264; CD45 associated protein; CD-45-AP; CD45-AP; CD45-associated protein; LPAP; LSM-1; Lymphocyte phosphatase-associated phosphoprotein; protein tyrosine phosphatase receptor type C-associated protein; protein tyrosine phosphatase, receptor type C associated protein; protein tyrosine phosphatase, receptor type, C polypeptide-associated protein; protein tyrosine phosphatase, receptor type, C-associated protein; Ptprcap; PTPRC-associated protein | |
| PTPRCAP | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human PTPRCAP (aa 112-169) Control Fragment | |
| RUO | |
| PTPRCAP | |
| Unconjugated | |
| Recombinant | |
| DTDYDHVADGGLQADPGEGEQQCGEASSPEQVPVRAEEARDSDTEGDLVLGSPGPASA | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |