missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PTPN20A (aa 60-87) Control Fragment Recombinant Protein

Catalog No. RP104358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104358 Supplier Invitrogen™ Supplier No. RP104358
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60332 (PA5-60332. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Tyrosine-protein phosphatase targeted to sites of actin polymerization in response of varied extracellular stimuli. Has tyrosine phosphatase activity towards various tyrosyl phosphorylated substrates.

Specifications

Accession Number Q4JDL3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26095
Name Human PTPN20A (aa 60-87) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias bA142I17.1; bA42B19.1; CT126; hPTPN20; protein tyrosine phosphatase, non-receptor type 20; protein tyrosine phosphatase, non-receptor type 20 A; protein tyrosine phosphatase, non-receptor type 20 B; PTPN20; PTPN20A; PTPN20B; Tyrosine-protein phosphatase non-receptor type 20
Common Name PTPN20A
Gene Symbol PTPN20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NFPHNDYEDVFEEPSESGSDPSMWTARG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less