missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRUNE2 Control Fragment Recombinant Protein

Catalog No. RP93020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93020 Supplier Invitrogen™ Supplier No. RP93020

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56632 (PA5-56632. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PRUNE2 gene, also named BMCC1, encodes protein prune homolog 2. PRUNE2 gene are overlapping with PCA3, one of the most prostate cancer specific markers. PRUNE2 may play an important role in regulating differentiation, survival and aggressiveness of the tumor cells. Besides its high level of expression seen in the nervous system (brain, cerebellum and spinal cord), it is also expressed at high levels in noneuroblastoma, rhabdomyosarcoma, melanoma and some osteosarcoma cell lines, whereas at only low levels in cancer cell lines of liver, breast, thyroid and colon. The Calculated molecular weight of this protein is 340 kDa.

Specifications

Accession Number Q8WUY3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 158471
Name Human PRUNE2 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330414G02Rik; A214N16.3; A230083H22Rik; A330102H22Rik; bA214N16.3; BCH motif-containing molecule at the carboxyl terminal region 1; Bmcc1; BNIP2 motif containing molecule at the carboxyl terminal region 1; BNIP2 motif-containing molecule at the C-terminal region 1; BNIPXL; C9orf65; DKFZp762K117; KIAA0367; mKIAA0367; neuronal protein; olfaxin; protein prune homolog 2; prune homolog 2; prune homolog 2 (Drosophila); PRUNE2; RGD1311350; RP11-214N16.3; RP11-58J3.2; truncated PRUNE2
Common Name PRUNE2
Gene Symbol PRUNE2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MESEKISEKQEEILSILEEKFPNLPPREDIINVLQETQFSAQGLSIEQTMLKDLKELSDGEIKVAISTVSMNLEVRVGML
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less