missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRMT9 (aa 233-331) Control Fragment Recombinant Protein

Catalog No. RP96723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96723 Supplier Invitrogen™ Supplier No. RP96723

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57845 (PA5-57845. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins, such as DTL/CDT2, BCL6 and PRDM1/BLIMP1. The SCF(FBXO11) complex mediates ubiquitination and degradation of BCL6, thereby playing a role in the germinal center B-cells terminal differentiation toward memory B-cells and plasma cells. The SCF(FBXO11) complex also mediates ubiquitination and degradation of DTL, an important step for the regulation of TGF-beta signaling, cell migration and the timing of the cell-cycle progression and exit. Binds to and neddylates phosphorylated p53/TP53, inhibiting its transcriptional activity. SCF(FBXO11) does not seem to direct ubiquitination of p53/TP53.

Specifications

Accession Number Q6P2P2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 90826
Name Human PRMT9 (aa 233-331) Control Fragment
Quantity 100 μL
Gene Alias AI931714; Prmt10; PRMT9; protein arginine methyltransferase 10 (putative); protein arginine methyltransferase 9; Protein arginine N-methyltransferase 10; Protein arginine N-methyltransferase 9; putative protein arginine N-methyltransferase 10; putative protein arginine N-methyltransferase 9; RGD1306157
Common Name PRMT9
Gene Symbol PRMT9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HTKSLDIEIPKHIPERVSLVVTETVDAGLFGEGIVESLIHAWEHLLLQPKTKGESANCEKYGKVIPASAVIFGMAVECAEIRRHHRVGIKDIAGIHLPT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less