missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PPAP2C (aa 252-287) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP91215
Description
Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84259 (PA5-84259. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.Specifications
| O43688 | |
| Blocking Assay, Control | |
| 8612 | |
| 100 μL | |
| lipid phosphate phosphohydrolase 2; LPP2; LPR3; PAP2c; PAP-2 c; PAP2-G; PAP2-gamma; phosphatidate phosphohydrolase type 2 c; Phosphatidic acid phosphatase 2 c; phosphatidic acid phosphatase type 2 C; phosphatidic acid phosphohydrolase type 2 c; phospholipid phosphatase 2; PLPP2; Ppap2c; PRG2; PRG-2; type-2 phosphatidic acid phosphatase-gamma | |
| PLPP2 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human PPAP2C (aa 252-287) Control Fragment | |
| RUO | |
| PPAP2C | |
| Unconjugated | |
| Recombinant | |
| PQHCLKEEELERKPSLSLTLTLGEADHNHYGYPHSS | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |