missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PP14B (aa 92-118) Control Fragment Recombinant Protein

Catalog No. RP101495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101495 Supplier Invitrogen™ Supplier No. RP101495

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62861 (PA5-62861. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.

Specifications

Accession Number Q96C90
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26472
Name Human PP14B (aa 92-118) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AOM172; Phi1; PHI-1; Phosphatase holoenzyme inhibitor 1; phospholipase C beta-3 neighboring gene protein; phospholipase c neighboring; phospholipase C-beta-3 neighbouring gene protein; PLCB3N; PNG; Ppp1r14b; protein phosphatase 1 regulatory inhibitor subunit 14 B; protein phosphatase 1 regulatory subunit 14 B; protein phosphatase 1, regulatory (inhibitor) 5 subunit 14 B; protein phosphatase 1, regulatory (inhibitor) subunit 14 B; SOM172; ubiquitous PKC-potentiated PP1 inhibitor
Common Name PP14B
Gene Symbol PPP1R14B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ELEIDVDELLDMESDDARAARVKELLV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less