missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human PORCN (NP_982301.1) VersaClone cDNA

Specifications
Specifications
| Gene | PORCN |
| Accession Number | NP_982301.1 |
| Gene ID (Entrez) | 64840 |
| Gene Alias | 2410004O13Rik, DHOF, EC 2.3.1.-, EC 2.3.1.250, FODH, MG61, MG61PPNprobable protein-cysteine N-palmitoyltransferase porcupine, por, PORC, PORCMGC29687, porcupine homolog (Drosophila), PPN, Protein MG61 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |