missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human POPDC2 (aa 204-324) Control Fragment Recombinant Protein

Catalog No. RP90944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP90944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP90944 Supplier Invitrogen™ Supplier No. RP90944

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55029 (PA5-55029. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the POP family of proteins which contain three putative transmembrane domains. This membrane associated protein is predominantly expressed in skeletal and cardiac muscle, and may have an important function in these tissues.

Specifications

Accession Number Q9HBU9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64091
Name Human POPDC2 (aa 204-324) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AV006127; Pop2; POPDC2; popeye 2; popeye domain containing 2; popeye domain-containing 2; Popeye domain-containing protein 2; popeye protein 2
Common Name POPDC2
Gene Symbol POPDC2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TAETSCSYISWPRKSLHLLLTKERYISCLFSALLGYDISEKLYTLNDKLFAKFGLRFDIRLPSLYHVLGPTAADAGPESEKGDEEVCEPAVSPPQATPTSLQQTPPCSTPPATTNFPAPPT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less